missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethylbiphenyl 99.0+%, TCI America™

Catalog No. H060825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H060825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H060825G Supplier TCI America Supplier No. H060825G

Chemical Identifiers

CAS 3597-91-9
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004660
InChI Key AXCHZLOJGKSWLV-UHFFFAOYSA-N
Synonym 4-biphenylmethanol, 4-phenylphenyl methanol, biphenyl-4-ylmethanol, 4-phenylbenzyl alcohol, p-phenylbenzyl alcohol, 4-hydroxymethylbiphenyl, 1,1'-biphenyl-4-ylmethanol, biphenyl-4-methanol, 4-hydroxymethyl biphenyl, 4hmb
PubChem CID 19186
IUPAC Name (4-phenylphenyl)methanol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CO

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxymethylbiphenyl
RTECSNumber DV5425000
TSCA No