missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethylbenzoic Acid 98.0+%, TCI America™

Catalog No. H076910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H076910G 10 g
H076925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H076910G Supplier TCI America Supplier No. H076910G

Chemical Identifiers

CAS 3006-96-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00017598
InChI Key WWYFPDXEIFBNKE-UHFFFAOYSA-N
Synonym 4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
PubChem CID 76360
IUPAC Name 4-(hydroxymethyl)benzoic acid
SMILES OCC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 183°C
Color White
Quantity 10 g
Formula Weight 152.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxymethylbenzoic Acid