missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H120425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H120425G 25 g
H12045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H120425G Supplier TCI America Supplier No. H120425G

Specifications

CAS 59016-93-2
Color White-Yellow
Molecular Formula C7H9BO3
MDL Number MFCD00792672
Quantity 25 g
Synonym 4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid
InChI Key PZRPBPMLSSNFOM-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)CO)(O)O
IUPAC Name [4-(hydroxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 151.956
PubChem CID 2734706
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less