missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenoxyacetic Acid 98.0+%, TCI America™

Catalog No. h09611g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09611G Supplier TCI America Supplier No. H09611G
Only null left

Chemical Identifiers

CAS 68858-21-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00057827
InChI Key VUCNQOPCYRJCGQ-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenoxyacetic acid, 4-hydroxymethylphenoxyacetic acid, 2-4-hydroxymethyl phenoxy acetic acid, hmp linker, p-hydroxymethyl phenoxyacetic acid, 4-hydroxymethyl-phenoxy acetic acid
PubChem CID 194291
IUPAC Name 2-[4-(hydroxymethyl)phenoxy]acetic acid
SMILES C1=CC(=CC=C1CO)OCC(=O)O

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymethyl)phenoxyacetic Acid