missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7130942
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12575G 5 g
H125725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12575G Supplier TCI America Supplier No. H12575G

Chemical Identifiers

CAS 33893-85-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
InChI Key VGRZISGVNOKTQU-UHFFFAOYSA-N
PubChem CID 12892866
IUPAC Name 4-(hydroxymethyl)cyclohexan-1-ol
SMILES C1CC(CCC1CO)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture)