Learn More
4-(Hydroxymethyl)benzeneboronic acid pinacol ester
CAS: 302348-51-2 | C13H19BO3 | 234.102 g/mol
Supplier: Thermo Scientific Chemicals H5112306
Description
4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 4-(Hydroxymethyl)benzeneboronic acid pinacol ester | |
| C13H19BO3 | |
| 5 g | |
| Insoluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CO | |
| 234.102 | |
| 234.1 |
| 302348-51-2 | |
| MFCD09837617 | |
| 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethyl phenylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethylphenylboronic acid pinacol ester, 4-hydroxymethyl benzeneboronic acid pinacol ester, 2-4-hydroxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzenemethanol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl alcohol, amtb135 | |
| GZZBZWITJNATOD-UHFFFAOYSA-N | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol | |
| 11402050 |
Chemical Identifiers
| 302348-51-2 | |
| 234.102 | |
| GZZBZWITJNATOD-UHFFFAOYSA-N | |
| 11402050 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CO |
| C13H19BO3 | |
| MFCD09837617 | |
| 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethyl phenylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethylphenylboronic acid pinacol ester, 4-hydroxymethyl benzeneboronic acid pinacol ester, 2-4-hydroxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzenemethanol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl alcohol, amtb135 | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only