missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzeneboronic acid pinacol ester

Catalog No. AAH5112314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5112314 25 g
AAH5112303 1 g
AAH5112306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5112314 Supplier Thermo Scientific Chemicals Supplier No. H5112314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 302348-51-2 | C13H19BO3 | 234.102 g/mol

4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 302348-51-2
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.102
MDL Number MFCD09837617
InChI Key GZZBZWITJNATOD-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethyl phenylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethylphenylboronic acid pinacol ester, 4-hydroxymethyl benzeneboronic acid pinacol ester, 2-4-hydroxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzenemethanol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl alcohol, amtb135
PubChem CID 11402050
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CO

Specifications

Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 234.1
Chemical Name or Material 4-(Hydroxymethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only