missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxychalcone 95.0+%, TCI America™

Catalog No. H09455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09455G Supplier TCI America Supplier No. H09455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2657-25-2
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016484
InChI Key UAHGNXFYLAJDIN-IZZDOVSWSA-N
Synonym 4'-hydroxychalcone, chalcone, 4'-hydroxy, 1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, 2e-1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, p-cinnamoylphenol, 2-benzal-4'-hydroxyacetophenone, hydroxychalcone, 4', 2-propen-1-one, 1-4-hydroxyphenyl-3-phenyl, unii-2k338k8uoa, e-1-4-hydroxyphenyl-3-phenyl-prop-2-en-1-one
PubChem CID 5282362
ChEBI CHEBI:34360
IUPAC Name (E)-1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 174°C
Color Yellow
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxychalcone