accessibility menu, dialog, popup

UserName

CAS RN 2657-25-2

IUPAC Name: (2E)-1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one
Synonyms: 4'-hydroxychalcone
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: UAHGNXFYLAJDIN-IZZDOVSWSA-N
SMILES: OC1=CC=C(C=C1)C(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)