missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxychalcone, 97%

Catalog No. AAA1083806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1083806 5 g
AAA1083814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1083806 Supplier Thermo Scientific Chemicals Supplier No. A1083806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2657-25-2 | C15H12O2 | 224.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2657-25-2
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016484
InChI Key UAHGNXFYLAJDIN-IZZDOVSWSA-N
Synonym 4'-hydroxychalcone, chalcone, 4'-hydroxy, 1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, 2e-1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, p-cinnamoylphenol, 2-benzal-4'-hydroxyacetophenone, hydroxychalcone, 4', 2-propen-1-one, 1-4-hydroxyphenyl-3-phenyl, unii-2k338k8uoa, e-1-4-hydroxyphenyl-3-phenyl-prop-2-en-1-one
PubChem CID 5282362
ChEBI CHEBI:34360
IUPAC Name (E)-1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 175°C to 177°C
Quantity 5 g
Beilstein 2049150
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material 4'-Hydroxychalcone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only