missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycarbazole 98.0+%, TCI America™

Catalog No. H10141G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10141G 1 g
H10145G 5 g
H101425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H10141G Supplier TCI America Supplier No. H10141G

Chemical Identifiers

CAS 52602-39-8
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD02178385
InChI Key UEOHATPGKDSULR-UHFFFAOYSA-N
Synonym 4-hydroxycarbazole, 4-hydroxy carbazole, ccris 5300, chembl46723, 4-hydroxycarbazol, 4-hydroxy-carbazole, pubchem9190, 4-hydroxy-9h-carbazole, acmc-209l0g
PubChem CID 104251
IUPAC Name 9H-carbazol-4-ol
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC=C3O

Specifications

Melting Point 169°C
Quantity 1 g
Formula Weight 183.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxycarbazole