accessibility menu, dialog, popup

UserName

OHHN

CAS RN 52602-39-8

IUPAC Name: 9H-carbazol-4-ol
Synonyms: 9H-carbazol-4-ol
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: UEOHATPGKDSULR-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C1=C(N2)C=CC=C1
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)