missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzylideneacetone, 97%

Catalog No. p-7023745
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1760514 25 g
AAA1760522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1760514 Supplier Thermo Scientific Chemicals Supplier No. A1760514
Only null left

CAS: 3160-35-8 | C10H10O2 | 162.19 g/mol

4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

Solubility
Optimally soluble in aqueous solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.

Chemical Identifiers

CAS 3160-35-8
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00016490
InChI Key OCNIKEFATSKIBE-NSCUHMNNSA-N
Synonym p-hydroxybenzalacetone, 4-hydroxybenzylideneacetone, 4-hydroxybenzal acetone, 4-hydroxycinnamoylmethane, 4-hydroxybenzalacetone, p-hydroxybenzylidene acetone, 4-p-hydroxyphenyl-3-buten-2-one, 4-4-hydroxyphenyl but-3-en-2-one, 3-buten-2-one, 4-4-hydroxyphenyl, 3-buten-2-one, 4-p-hydroxyphenyl
PubChem CID 796857
SMILES CC(=O)\C=C\C1=CC=C(O)C=C1

Specifications

Melting Point 106°C to 111°C
Quantity 25 g
Beilstein 2412544
Solubility Information Optimally soluble in aqueous solvents.
IUPAC Name (E)-4-(4-hydroxyphenyl)but-3-en-2-one
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzylideneacetone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-607-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only