missing translation for 'onlineSavingsMsg'
Learn More

(4-Hydroxybenzyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. H1558200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1558200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1558200MG Supplier TCI America Supplier No. H1558200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90001-07-3
Molecular Formula C7H9O4P
Molecular Weight (g/mol) 188.12
MDL Number MFCD16877729
InChI Key ZHBRSHSRMYZHLS-UHFFFAOYSA-N
PubChem CID 12029255
IUPAC Name [(4-hydroxyphenyl)methyl]phosphonic acid
SMILES OC1=CC=C(CP(O)(O)=O)C=C1

Specifications

Melting Point 222°C
Quantity 200 mg
UN Number 3261
Formula Weight 188.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Hydroxybenzyl)phosphonic Acid