missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl Alcohol 97.0+%, TCI America™

Catalog No. p-7130422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022425G 25 g
H0224100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022425G Supplier TCI America Supplier No. H022425G

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004658
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzyl Alcohol
EINECSNumber (3)-1052
TSCA No
Recommended Storage Refrigerator