missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl alcohol, 99%

Catalog No. p-7020985
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1313218 50 g
AAA1313222 100 g
AAA1313230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1313218 Supplier Thermo Scientific Chemicals Supplier No. A1313218

CAS: 623-05-2 | C7H8O2 | 124.139 g/mol

4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004658
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O

Specifications

Melting Point 114°C to 122°C
Boiling Point 251°C to 253°C
Quantity 50 g
Beilstein 1858967
Sensitivity Air and light sensitive
Solubility Information Soluble in water (6.7mg/ml at 20°C),dioxane (100mg/ml),1N NaOH (50mg/ml),DMSO,and methanol.
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 4-Hydroxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-768-0
RTECSNumber DA4796800
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only