missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone, 98+%, Thermo Scientific™

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002355
InChI Key NPFYZDNDJHZQKY-UHFFFAOYSA-N
Synonym 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
PubChem CID 14347
ChEBI CHEBI:34421
IUPAC Name (4-hydroxyphenyl)-phenylmethanone
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121010250
SDS View Documents
Thermo Scientific Chemicals
121010250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC121011000
SDS View Documents
Thermo Scientific Chemicals
121011000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1137-42-4
198.22
NPFYZDNDJHZQKY-UHFFFAOYSA-N
14347
(4-hydroxyphenyl)-phenylmethanone
C13H10O2
MFCD00002355
4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
CHEBI:34421
OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

1137-42-4
100.0
98% min. (GC)
C13H10O2
MFCD00002355
08,II,184
Solubility in water: insoluble
OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
198.22
CHEBI:34421
98+%
98.0
Authentic
Glass bottle
HOC6H4COC6H5
25g
4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
NPFYZDNDJHZQKY-UHFFFAOYSA-N
(4-hydroxyphenyl)-phenylmethanone
14347
198.22
4-Hydroxybenzophenone, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-507-1

RTECSNumber : PC4959775

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only