missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzonitrile, 98+%

Catalog No. p-7022852
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1580318 50 g
AAA1580322 100 g
AAA1580330 250 g
AAA1580336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1580318 Supplier Thermo Scientific Chemicals Supplier No. A1580318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 767-00-0 | C7H5NO | 119.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 767-00-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002312
InChI Key CVNOWLNNPYYEOH-UHFFFAOYSA-N
Synonym 4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
PubChem CID 13019
ChEBI CHEBI:38622
IUPAC Name 4-hydroxybenzonitrile
SMILES C1=CC(=CC=C1C#N)O

Specifications

Melting Point 110°C to 114°C
Quantity 50 g
Beilstein 386130
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 119.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxybenzonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-175-2
RTECSNumber DI4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only