missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzohydrazide 98.0+%, TCI America™

Catalog No. p-7130955
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H127525G 25 g
H1275100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H127525G Supplier TCI America Supplier No. H127525G
Only null left

Chemical Identifiers

CAS 5351-23-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007605
InChI Key ZMZGIVVRBMFZSG-UHFFFAOYSA-N
Synonym 4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide
PubChem CID 1742
IUPAC Name 4-hydroxybenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)O

Specifications

Melting Point 268°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzohydrazide