missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzhydrazide, 98%

Catalog No. AAA1270214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1270214 25 g
AAA1270222 100 g
AAA1270236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1270214 Supplier Thermo Scientific Chemicals Supplier No. A1270214

CAS: 5351-23-5 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5351-23-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007605
InChI Key ZMZGIVVRBMFZSG-UHFFFAOYSA-N
Synonym 4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide
PubChem CID 1742
IUPAC Name 4-hydroxybenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)O

Specifications

Melting Point ∼267°C (decomposition)
Quantity 25 g
Beilstein 473052
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-326-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only