missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenethiol 97.0+%, TCI America™

Catalog No. H066225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H066225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H066225G Supplier TCI America Supplier No. H066225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-89-8
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00004850
InChI Key BXAVKNRWVKUTLY-UHFFFAOYSA-N
Synonym 4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio
PubChem CID 240147
IUPAC Name 4-sulfanylphenol
SMILES OC1=CC=C(S)C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 126.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Hydroxybenzenethiol
RTECSNumber MX7175000
TSCA No