missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyazobenzene-4-carboxylic Acid Hydrate 98.0+%, TCI America™

Catalog No. H1400200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1400200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1400200MG Supplier TCI America Supplier No. H1400200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2497-38-3
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD12198372
InChI Key XLEOLQBYKOFJPH-UHFFFAOYSA-N
Synonym 4-(4-Hydroxyphenylazo)benzoic Acid
PubChem CID 5371178
IUPAC Name 4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
SMILES C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 260°C
Quantity 200 mg
Formula Weight 242.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyazobenzene-4-carboxylic Acid Hydrate