accessibility menu, dialog, popup

UserName

CAS RN 2497-38-3

IUPAC Name: 4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzoic acid
Synonyms: 4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzoic acid
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: XLEOLQBYKOFJPH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NN=C2C=CC(=O)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results