missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxyacetophenone 98.0+%, TCI America™
$55.84 - $398.51
Chemical Identifiers
| CAS | 99-93-4 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00002359 |
| InChI Key | TXFPEBPIARQUIG-UHFFFAOYSA-N |
| Synonym | 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone |
| PubChem CID | 7469 |
| ChEBI | CHEBI:28032 |
| IUPAC Name | 1-(4-hydroxyphenyl)ethan-1-one |
| SMILES | CC(=O)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 99-93-4 | |
| 136.15 | |
| TXFPEBPIARQUIG-UHFFFAOYSA-N | |
| 7469 | |
| 1-(4-hydroxyphenyl)ethan-1-one |
| C8H8O2 | |
| MFCD00002359 | |
| 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
| CHEBI:28032 | |
| CC(=O)C1=CC=C(O)C=C1 |
Specifications
| 99-93-4 | |
| White | |
| C8H8O2 | |
| 25 g | |
| TXFPEBPIARQUIG-UHFFFAOYSA-N | |
| 1-(4-hydroxyphenyl)ethan-1-one | |
| 7469 | |
| 136.15 | |
| Crystalline Powder |
| 110°C | |
| 148°C | |
| MFCD00002359 | |
| 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
| CC(=O)C1=CC=C(O)C=C1 | |
| 136.15 | |
| CHEBI:28032 | |
| ≥98.0% (HPLC,T) | |
| 4′-Hydroxyacetophenone |
Safety and Handling
EINECSNumber : (3)-4119
RTECSNumber : AM8750000
TSCA : Yes