missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxyacetophenone 98.0+%, TCI America™
Supplier: TCI America H019325G
Specifications
| 4′-Hydroxyacetophenone | |
| 110°C | |
| 148°C | |
| MFCD00002359 | |
| 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
| CC(=O)C1=CC=C(O)C=C1 | |
| 136.15 | |
| CHEBI:28032 | |
| ≥98.0% (HPLC,T) |
| 99-93-4 | |
| White | |
| C8H8O2 | |
| 25 g | |
| TXFPEBPIARQUIG-UHFFFAOYSA-N | |
| 1-(4-hydroxyphenyl)ethan-1-one | |
| 7469 | |
| 136.15 | |
| Crystalline Powder |
Chemical Identifiers
| 99-93-4 | |
| 136.15 | |
| TXFPEBPIARQUIG-UHFFFAOYSA-N | |
| 7469 | |
| 1-(4-hydroxyphenyl)ethan-1-one |
| C8H8O2 | |
| MFCD00002359 | |
| 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
| CHEBI:28032 | |
| CC(=O)C1=CC=C(O)C=C1 |
Safety and Handling
EINECSNumber : (3)-4119
RTECSNumber : AM8750000
TSCA : Yes