missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone Oxime 98.0+%, TCI America™

Catalog No. H15121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15121G 1 g
H15125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15121G Supplier TCI America Supplier No. H15121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34523-34-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00481697
InChI Key NOGIFJNJJZFUDN-UHFFFAOYSA-N
PubChem CID 5357461
IUPAC Name 4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one
SMILES CC(NO)=C1C=CC(=O)C=C1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyacetophenone Oxime
TSCA Yes
Recommended Storage Refrigerator