accessibility menu, dialog, popup

UserName

CAS RN 34523-34-7

IUPAC Name: 4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one
Synonyms: 4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: NOGIFJNJJZFUDN-UHFFFAOYSA-N
SMILES: CC(NO)=C1C=CC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results