missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyacetophenone, Analytical Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111016028
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016028 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016028 Supplier Merck Emd Millipore Supplier No. 98837100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 99-93-4
Physical Form Neat
Linear Formula HOC6H4COCH3
MDL Number MFCD00002359
Melting Point 107°C to 110°C; 109°C to 111°C
Boiling Point 147°C to 148°C (3 mmHg)
Synonym 4-Hydroxyphenylethanone; p-Acetophenol; p-Hydroxyphenyl methyl ketone; Piceol
Quantity 100 mg
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less