missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetanilide 98.0+%, TCI America™

Catalog No. p-7130394
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H019025G 25 g
H0190500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. H019025G missing translation for 'mfr' TCI America missing translation for 'supplierNo' H019025G
Only null left

Chemical Identifiers

CAS 103-90-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00002328
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
PubChem CID 1983
ChEBI CHEBI:46195
IUPAC Name N-(4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C=C1

Specifications

Melting Point 169°C
Color White
pH 5.5 to 6.5 (saturated H2O soln.)
Quantity 25 g
UN Number 3077
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyacetanilide
EINECSNumber (3)-0678
RTECSNumber AE4200000
TSCA Yes