missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-L-(+)-2-phenylglycine 99.0+%, TCI America™
$180.81 - $544.30
Chemical Identifiers
| CAS | 32462-30-9 |
|---|---|
| Molecular Formula | C8H9NO3 |
| Molecular Weight (g/mol) | 167.164 |
| MDL Number | MFCD00065932 |
| InChI Key | LJCWONGJFPCTTL-ZETCQYMHSA-N |
| Synonym | (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH |
| PubChem CID | 36143 |
| ChEBI | CHEBI:31755 |
| IUPAC Name | (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid |
| SMILES | C1=CC(=CC=C1C(C(=O)O)N)O |
Chemical Identifiers
| 32462-30-9 | |
| 167.164 | |
| LJCWONGJFPCTTL-ZETCQYMHSA-N | |
| 36143 | |
| (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid |
| C8H9NO3 | |
| MFCD00065932 | |
| (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH | |
| CHEBI:31755 | |
| C1=CC(=CC=C1C(C(=O)O)N)O |
Specifications
| 32462-30-9 | |
| White-Yellow | |
| MFCD00065932 | |
| (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH | |
| C1=CC(=CC=C1C(C(=O)O)N)O | |
| 167.164 | |
| CHEBI:31755 | |
| ≥99.0% (HPLC,T) | |
| 4-Hydroxy-L-(+)-2-phenylglycine |
| 214°C | |
| C8H9NO3 | |
| 5 g | |
| LJCWONGJFPCTTL-ZETCQYMHSA-N | |
| (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid | |
| 36143 | |
| 167.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3667
TSCA : Yes