missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-D-(-)-2-phenylglycine 99.0+%, TCI America™

Catalog No. H0758500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0758500G 500 g
H075825G 25 g
H0758100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0758500G Supplier TCI America Supplier No. H0758500G

Chemical Identifiers

CAS 22818-40-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00004262
InChI Key LJCWONGJFPCTTL-SSDOTTSWSA-N
Synonym d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
PubChem CID 89853
ChEBI CHEBI:15695
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 500 g
Formula Weight 167.16
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-D-(-)-2-phenylglycine
EINECSNumber (9)-1624
TSCA Yes