missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-D-(-)-2-phenylglycine 99.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 240°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 167.16 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Hydroxy-D-(-)-2-phenylglycine |
Sécurité et manipulation
| EINECSNumber | (9)-1624 |
| TSCA | Yes |