missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-7-methoxycoumarin 98.0+%, TCI America™

Catalog No. H14201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14201G 1 g
H14205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14201G Supplier TCI America Supplier No. H14201G

Chemical Identifiers

CAS 17575-15-4
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00673700
InChI Key HFGKBQZAHZKDLV-UHFFFAOYSA-N
Synonym 4-hydroxy-7-methoxycoumarin, 4-hydroxy-7-methoxy-2h-chromen-2-one, 2-hydroxy-7-methoxy-4h-chromen-4-one, 2-hydroxy-7-methoxychromen-4-one, 4-hydroxy-7-methoxy-2h-1-benzopyran-2-one, 4-hydroxy-7-met hoxycoumarin, 4-hydroxy-7-methoxy-coumarin, 4-hydroxy-7-methoxy-2-oxo-2h-chromene, 2h-1-benzopyran-2-one,4-hydroxy-7-methoxy, 2h-1-benzopyran-2-one, 4-hydroxy-7-methoxy
PubChem CID 54691408
IUPAC Name 2-hydroxy-7-methoxy-4H-chromen-4-one
SMILES COC1=CC=C2C(=O)C=C(O)OC2=C1

Specifications

Melting Point 260°C
Quantity 1 g
Formula Weight 192.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-7-methoxycoumarin