accessibility menu, dialog, popup

UserName

CAS RN 17575-15-4

IUPAC Name: 2-hydroxy-7-methoxy-4H-chromen-4-one
Synonyms: 2-hydroxy-7-methoxychromen-4-one
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: HFGKBQZAHZKDLV-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=O)C=C(O)OC2=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)