missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-nicotinic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6649906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6649906 5 g
AAH6649903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6649906 Supplier Thermo Scientific Chemicals Supplier No. H6649906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33821-58-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD04038153
InChI Key AOJLDZLRTUWFFY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methylnicotinic acid, 6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 4-hydroxy-6-methyl-nicotinic acid, 4-hydroxy 6-methylnicotinic acid, 4-hydroxy-6-methylpyridine-3-carboxylic acid, 4-hydroxy-6-methyl-3-pyridinecarboxylic acid, 6-methyl-4-hydroxynicotinic acid, chembrdg-bb 4002882, 6-methyl-4 1h-pyridone-3-carboxylic acid, 4-hydroxy-6-methyl-3-picolinic acid
PubChem CID 10702014
IUPAC Name 6-methyl-4-oxo-1H-pyridine-3-carboxylic acid
SMILES CC1=CC(=O)C(=CN1)C(=O)O

Specifications

Quantity 5 g
Beilstein 387402
Formula Weight 153.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-6-methyl-nicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only