accessibility menu, dialog, popup

UserName

CAS RN 33821-58-8

IUPAC Name: 6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid
Synonyms: 6-methyl-4-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: AOJLDZLRTUWFFY-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C(=CN1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results