missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-3-nitro-2-pyridone 98.0+%, TCI America™

Catalog No. p-7130808
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
H09215G 5 g
H092125G 25 g
2 options

Catalog No. H09215G

Supplier: TCI America H09215G

Only null left

Chemical Identifiers

CAS 4966-90-9
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00167612
InChI Key QIKWTNPFTOEELW-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, 2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, pubchem3892, acmc-209khe
PubChem CID 54685619
IUPAC Name 2-hydroxy-6-methyl-3-nitro-1,4-dihydropyridin-4-one
SMILES CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 170.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-6-methyl-3-nitro-2-pyridone