accessibility menu, dialog, popup

UserName

CAS RN 4966-90-9

IUPAC Name: 2-hydroxy-6-methyl-3-nitro-1,4-dihydropyridin-4-one
Synonyms: 2-hydroxy-6-methyl-3-nitro-1H-pyridin-4-one
Molecular Weight (g/mol): 170.12
Molecular Formula: C6H6N2O4
InChi Key: QIKWTNPFTOEELW-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)