missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-nitrostilbene, 98%

Catalog No. p-7025443
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2091303 1 g
AAB2091306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2091303 Supplier Thermo Scientific Chemicals Supplier No. B2091303

CAS: 19221-08-0 | C14H11NO3 | 241.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19221-08-0
Molecular Formula C14H11NO3
Molecular Weight (g/mol) 241.246
MDL Number MFCD00017044
InChI Key OETQWIHJPIESQB-OWOJBTEDSA-N
Synonym 4-hydroxy-4'-nitrostilbene, 4-4-nitrostyryl phenol, 4-e-2-4-nitrophenyl ethenyl phenol, 4-hydroxy-4/'-nitrostilbene, 4-1e-2-4-nitrophenyl vinyl phenol, 4'-nitrostilben-4-ol, cambridge id 5309411, e-4'-nitrostilbene-4-ol, e-4-4-nitrostyryl phenol
PubChem CID 759250
IUPAC Name 4-[(E)-2-(4-nitrophenyl)ethenyl]phenol
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 205°C to 209°C
Quantity 1 g
Beilstein 2505854
Formula Weight 241.25
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-4'-nitrostilbene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only