missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-methyl-2-pentanone, 99%

Catalog No. p-4687109
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121482500 250 mL Glass bottle
AC121480010 1 L Glass bottle
AC121480025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121482500 Supplier Thermo Scientific Chemicals Supplier No. 121482500
Only null left

CAS: 123-42-2 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-42-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004471
InChI Key SWXVUIWOUIDPGS-UHFFFAOYSA-N
Synonym diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
PubChem CID 31256
ChEBI CHEBI:55381
IUPAC Name 4-hydroxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)O

Specifications

Melting Point -42.8°C
Color Colorless
Density 0.9385g/mL
Boiling Point 166.0°C
Flash Point 58°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4223 to 1.4243
Linear Formula (CH3)2C(OH)CH2COCH3
Quantity 250 mL
Merck Index 15,2967
Specific Gravity 0.9385
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,ether
Viscosity 3.22 mPa.s (20°C)
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-4-methyl-2-pentanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye
EINECSNumber 204-626-7

RUO – Research Use Only