missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-methoxybiphenyl 95.0+%, TCI America™

Catalog No. H08191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08191G 1 g
H08195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08191G Supplier TCI America Supplier No. H08191G

Chemical Identifiers

CAS 16881-71-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00191514
InChI Key CORJIEYQXMZUIW-UHFFFAOYSA-N
PubChem CID 11030839
IUPAC Name 4-(4-methoxyphenyl)phenol
SMILES COC1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 181°C
Color White
Quantity 1 g
Synonym 4-Hydroxy-4′C-methoxydiphenyl, 4′C-Methoxybiphenyl-4-ol
Formula Weight 200.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-4′-methoxybiphenyl