missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-dimethylaminoazobenzene 98.0+%, TCI America™

Catalog No. H023725G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023725G 25 g
H02371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H023725G Supplier TCI America Supplier No. H023725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2496-15-3
Molecular Formula C14H15N3O
Molecular Weight (g/mol) 241.294
MDL Number MFCD00059624
InChI Key XBCKVYYJXKZISB-UHFFFAOYSA-N
Synonym 4-(4-Dimethylaminophenylazo)phenol
PubChem CID 5370977
IUPAC Name 4-[[4-(dimethylamino)phenyl]hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES CN(C)C1=CC=C(C=C1)NN=C2C=CC(=O)C=C2

Specifications

Melting Point 200°C
Quantity 25 g
Formula Weight 241.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-4′-dimethylaminoazobenzene