missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog No. p-7028693
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2609703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2609703 Supplier Thermo Scientific Chemicals Supplier No. H2609703
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 220239-68-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD01091008
InChI Key DPVRVZQEDJVWLS-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzoicacid, acmc-1cdpd, ksc495o6p, 4-hydroxy-3-trifluoromethyl-benzoic acid, 4-oxidanyl-3-trifluoromethyl benzoic acid
PubChem CID 2775112
IUPAC Name 4-hydroxy-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F

Specifications

Melting Point 144°C to 147°C
Quantity 1 g
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures