missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3'-(trifluoromethyl)acetophenone, 95%, Thermo Scientific Chemicals

Catalog No. AAH3169103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3169103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3169103 Supplier Thermo Scientific Chemicals Supplier No. H3169103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149105-11-3 | Molecular Formula: C9H7F3O2 | Molecular Weight: 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149105-11-3
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD01091005
InChI Key HKRUXZJKSFFSGF-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl acetophenone, 1-4-hydroxy-3-trifluoromethyl phenyl ethanone, 4'-hydroxy-3'-trifluoromethyl acetophenone, 1-4-hydroxy-3-trifluoromethyl phenyl ethan-1-one, 1-4-hydroxy-3-trifluoromethylphenyl ethanone, ethanone,1-4-hydroxy-3-trifluoromethyl phenyl, acmc-1bxyq, intermediates-zcf02143, 5-acetyl-2-hydroxybenzotrifluoride
PubChem CID 2775109
IUPAC Name 1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)C(F)(F)F

Specifications

Melting Point 174°C to 177°C
Quantity 1 g
Formula Weight 204.15
Percent Purity 95%
Chemical Name or Material 4'-Hydroxy-3'-(trifluoromethyl)acetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only