CAS RN 149105-11-3
IUPAC Name:
1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms:
1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol):
204.15
Molecular Formula:
C9H7F3O2
InChi Key:
HKRUXZJKSFFSGF-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(=C(O)C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4'-Hydroxy-3'-(trifluoromethyl)acetophenone, 95%, Thermo Scientific Chemicals
4'-Hydroxy-3'-(trifluoromethyl)acetophenone, 95%, Thermo Scientific Chemicals
CAS: 149105-11-3 Molecular Formula: C9H7F3O2 Molecular Weight (g/mol): 204.148 MDL Number: MFCD01091005 InChI Key: HKRUXZJKSFFSGF-UHFFFAOYSA-N Synonym: 4-hydroxy-3-trifluoromethyl acetophenone,1-4-hydroxy-3-trifluoromethyl phenyl ethanone,4'-hydroxy-3'-trifluoromethyl acetophenone,1-4-hydroxy-3-trifluoromethyl phenyl ethan-1-one,1-4-hydroxy-3-trifluoromethylphenyl ethanone,ethanone,1-4-hydroxy-3-trifluoromethyl phenyl,acmc-1bxyq,intermediates-zcf02143,5-acetyl-2-hydroxybenzotrifluoride PubChem CID: 2775109 IUPAC Name: 1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethanone SMILES: CC(=O)C1=CC(=C(C=C1)O)C(F)(F)F