missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3-tert-butylanisole 98.0+%, TCI America™
$59.69 - $394.66
Chemical Identifiers
| CAS | 121-00-6 |
|---|---|
| Molecular Formula | C11H16O2 |
| Molecular Weight (g/mol) | 180.247 |
| MDL Number | MFCD00040484 |
| InChI Key | MRBKEAMVRSLQPH-UHFFFAOYSA-N |
| Synonym | 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy |
| PubChem CID | 8456 |
| ChEBI | CHEBI:76358 |
| IUPAC Name | 2-tert-butyl-4-methoxyphenol |
| SMILES | CC(C)(C)C1=C(C=CC(=C1)OC)O |
Chemical Identifiers
| 121-00-6 | |
| 180.247 | |
| MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
| 8456 | |
| 2-tert-butyl-4-methoxyphenol |
| C11H16O2 | |
| MFCD00040484 | |
| 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
| CHEBI:76358 | |
| CC(C)(C)C1=C(C=CC(=C1)OC)O |
Specifications
| 121-00-6 | |
| C11H16O2 | |
| 25 g | |
| 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
| CC(C)(C)C1=C(C=CC(=C1)OC)O | |
| 180.247 | |
| CHEBI:76358 | |
| ≥98.0% (GC) | |
| 4-Hydroxy-3-tert-butylanisole |
| 60°C | |
| MFCD00040484 | |
| 3077 | |
| MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
| 2-tert-butyl-4-methoxyphenol | |
| 8456 | |
| 180.25 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0608&(9)-1199&(9)-1532
RTECSNumber : SK1575000
TSCA : Yes