missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitroso-1-naphthalenesulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. p-7133716
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02695G 5 g
N026925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02695G Supplier TCI America Supplier No. N02695G

Chemical Identifiers

CAS 3682-32-4
Molecular Formula C10H7NO5S
Molecular Weight (g/mol) 253.228
MDL Number MFCD00003933
InChI Key GASPSNIEUBWWJU-UHFFFAOYSA-N
Synonym 2-nitroso-1-naphthol-4-sulfonic acid, 4-hydroxy-3-nitroso-1-naphthalenesulfonic acid, 1-naphthalenesulfonic acid, 4-hydroxy-3-nitroso, unii-7t06pca43o, 4-hydroxy-3-nitroso-naphthalene-1-sulfonic acid, 4-hydroxy-3-nitrosonaphthalenesulfonic acid, nitroso-nw acid, acmc-1cril, 4-hydroxy-3-nitrosonaphthalene-1-sulphonic acid, 1-naphthalenesulfonicacid, 4-hydroxy-3-nitroso
PubChem CID 77257
IUPAC Name 4-hydroxy-3-nitrosonaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2O)N=O)S(=O)(=O)O

Specifications

Melting Point 145°C
Quantity 5 g
Formula Weight 253.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitroso-1-naphthalenesulfonic Acid Hydrate