missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7130802
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H091025G 25 g
H0910100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H091025G Supplier TCI America Supplier No. H091025G
Only null left

Chemical Identifiers

CAS 616-82-0
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007119
InChI Key QRYSWXFQLFLJTC-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxybenzoic acid, benzoic acid, 4-hydroxy-3-nitro, 3-nitro-4-hydrobenzoic acid, 4-hydroxy-3-nitro-benzoic acid, nhb, pubchem18981, acmc-1auqv, 3,4-nitrohydroxybenzoic acid, ksc489q0p
PubChem CID 12033
IUPAC Name 4-hydroxy-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O

Specifications

Melting Point 184°C
Quantity 25 g
Formula Weight 183.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrobenzoic Acid