missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzoic acid, 98%

Catalog No. AAA1056214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1056214 25 g
AAA1056222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1056214 Supplier Thermo Scientific Chemicals Supplier No. A1056214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-82-0 | C7H5NO5 | 183.12 g/mol

Standard in chymotrypsin assays

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-82-0
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007119
InChI Key QRYSWXFQLFLJTC-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxybenzoic acid, benzoic acid, 4-hydroxy-3-nitro, 3-nitro-4-hydrobenzoic acid, 4-hydroxy-3-nitro-benzoic acid, nhb, pubchem18981, acmc-1auqv, 3,4-nitrohydroxybenzoic acid, ksc489q0p
PubChem CID 12033
IUPAC Name 4-hydroxy-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O

Specifications

Melting Point 182°C to 184°C
Quantity 25 g
Beilstein 2213134
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-494-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only