missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3-methylbenzoic Acid 98.0+%, TCI America™
$107.80 - $431.25
Chemical Identifiers
| CAS | 499-76-3 |
|---|---|
| Molecular Formula | C8H8O3 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00270105 |
| InChI Key | LTFHNKUKQYVHDX-UHFFFAOYSA-N |
| Synonym | 4-hydroxy-m-toluic acid, 3-methyl-4-hydroxybenzoic acid, 4-hydroxy-3-methyl-benzoic acid, 4,3-cresotic acid, 4-hydroxy-3-methylbenzoate, benzoic acid, 4-hydroxy-3-methyl, 4-hydroxy-m-toluate, pubchem20044, acmc-209kj4, 3-methyl-4-hydroxybenzoate |
| PubChem CID | 68138 |
| ChEBI | CHEBI:85239 |
| IUPAC Name | 4-hydroxy-3-methylbenzoic acid |
| SMILES | CC1=CC(=CC=C1O)C(O)=O |
Chemical Identifiers
| 499-76-3 | |
| 152.15 | |
| LTFHNKUKQYVHDX-UHFFFAOYSA-N | |
| 68138 | |
| 4-hydroxy-3-methylbenzoic acid |
| C8H8O3 | |
| MFCD00270105 | |
| 4-hydroxy-m-toluic acid, 3-methyl-4-hydroxybenzoic acid, 4-hydroxy-3-methyl-benzoic acid, 4,3-cresotic acid, 4-hydroxy-3-methylbenzoate, benzoic acid, 4-hydroxy-3-methyl, 4-hydroxy-m-toluate, pubchem20044, acmc-209kj4, 3-methyl-4-hydroxybenzoate | |
| CHEBI:85239 | |
| CC1=CC(=CC=C1O)C(O)=O |
Specifications
| 499-76-3 | |
| C8H8O3 | |
| 5 g | |
| LTFHNKUKQYVHDX-UHFFFAOYSA-N | |
| 4-hydroxy-3-methylbenzoic acid | |
| 68138 | |
| 152.15 | |
| Crystalline Powder |
| 175°C | |
| MFCD00270105 | |
| 4-hydroxy-m-toluic acid, 3-methyl-4-hydroxybenzoic acid, 4-hydroxy-3-methyl-benzoic acid, 4,3-cresotic acid, 4-hydroxy-3-methylbenzoate, benzoic acid, 4-hydroxy-3-methyl, 4-hydroxy-m-toluate, pubchem20044, acmc-209kj4, 3-methyl-4-hydroxybenzoate | |
| CC1=CC(=CC=C1O)C(O)=O | |
| 152.15 | |
| CHEBI:85239 | |
| ≥98.0% (GC) | |
| 4-Hydroxy-3-methylbenzoic Acid |
Safety and Handling
TSCA : No